3-Iodphenylboronsäure ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

H51112.03 H51112.06
H51112.03EA 25.6 EUR
H51112.03 H51112.06
3-Iodphenylboronsäure ≥97%
3-Iodphenylboronsäure

Purity: 97% CAS No.: 221037-98-5 Molecular Formula: C6H6BIO2 Formula Weight: 247.83 MDL No.: MFCD01319015

Formel: C₆H₆BIO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01319015
CAS-Nummer: 221037-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR