5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
Lieferant: Thermo Scientific
Synonyme:
1-{[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl}piperidine
H51117.06
H51117.06EA
138
EUR
H51117.06
5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
5-(1-Piperidinylmethyl)thiophen-2-boronsäurepinakolester
CAS No.: 1218790-44-3
Molecular Formula: C16H26BNO2S
Formula Weight: 307.26
MDL No.: MFCD11113036
Formel:
C₁₆H₂₆BNO₂S Molecular Weight: 307,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11113036 CAS-Nummer: 1218790-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...