N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

Lieferant: Thermo Fisher Scientific

H51136.03 H51136.06 H51136.14
H51136.03EA 32.9 EUR
H51136.03 H51136.06 H51136.14
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine

Purity: 98% CAS No.: 62965-35-9 Molecular Formula: C11H21NO4 Formula Weight: 231.29 Melting Point: 118-121° MDL No.: MFCD00065574

Formel: C₁₁H₂₁NO₄
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065574
CAS-Nummer: 62965-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR