2-Butenylmagnesium chloride 0.5 M in MeTHF (2-Methyltetrahydrofuran)

Lieferant: Thermo Scientific
Gefahr

H51171.AE
H51171.AEEA 207 EUR
H51171.AE
2-Butenylmagnesium chloride 0.5 M in MeTHF (2-Methyltetrahydrofuran)
2-Butenylmagnesium chloride

CAS No.: 22649-70-3 Molecular Formula: C4H7ClMg Formula Weight: 114.86 UN No.: UN3399 MDL No.: MFCD01321226

Formel: C₄H₇ClMg
Molecular Weight: 114,86 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321226
CAS-Nummer: 6088-88-6
UN: 3399
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR