2,5-Diiod-1-methylimidazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,5-Diiod-1-methyl-1H-imidazol
H51212.03
H51212.03EA
226
EUR
H51212.03
2,5-Diiod-1-methylimidazol ≥98%
2,5-Diiodo-1-methylimidazole
Formel:
C₄H₄I₂N₂ Molecular Weight: 333,9 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179540 CAS-Nummer: 86026-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...