1-Methyl-5-(tributylstannyl)imidazol ≥90%
Lieferant: Thermo Scientific
|
Gefahr
|
H51405.03
H51405.03EA
178
EUR
H51405.03
1-Methyl-5-(tributylstannyl)imidazol ≥90%
1-Methyl-5-(tributylstannyl)imidazol
Purity: 90+%
CAS No.: 147716-03-8
Molecular Formula: C16H32N2Sn
Formula Weight: 371.15
UN No.: 2788
MDL No.: MFCD01319031
Formel:
C₁₆H₃₂N₂Sn Molecular Weight: 371,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319031 CAS-Nummer: 147716-03-8 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...