4-Methyl-2-(tributylstannyl)pyridin 96%
Lieferant: Thermo Scientific
|
Gefahr
|
H51472.03
H51472.03EA
171
EUR
H51472.03
4-Methyl-2-(tributylstannyl)pyridin 96%
4-Methyl-2-(tributylstannyl)pyridin
Purity: 96%
CAS No.: 301652-23-3
Molecular Formula: C18H33NSn
Formula Weight: 382.16
UN No.: 2788
MDL No.: MFCD07787403
Formel:
C₁₈H₃₃NSn Molecular Weight: 382,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787403 CAS-Nummer: 301652-23-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...