3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione
H51488.MD
H51488.MDEA
755
EUR
H51488.MD
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion ≥97%
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutendion
Purity: 97%
CAS No.: 129034-70-4
Molecular Formula: C19H34O3Sn
Formula Weight: 429.17
UN No.: 2788
MDL No.: MFCD01319011
Formel:
C₁₉H₃₄O₃Sn Molecular Weight: 429,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319011 CAS-Nummer: 129034-70-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...