1-Methyl-2-(tributylstannyl)imidazol ≥90%

Lieferant: Thermo Scientific
Gefahr

H51523.03
H51523.03EA 277 EUR
H51523.03
1-Methyl-2-(tributylstannyl)imidazol ≥90%
1-Methyl-2-(tributylstannyl)imidazol

Purity: 90+% CAS No.: 105494-69-7 Molecular Formula: C16H32N2Sn Formula Weight: 371.15 Storage Temperature: 2°C to 8°C

Formel: C₁₆H₃₂N₂Sn
Molecular Weight: 371,15 g/mol
MDL: MFCD01319030
CAS-Nummer: 105494-69-7
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR