2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin 96%
Lieferant: Thermo Scientific
|
Gefahr
|
H51574.03
H51574.03EA
420
EUR
H51574.03
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin 96%
2-(Methylsulfonyl)-5-(tributylstannyl)pyrimidin
Purity: 96%
CAS No.: 122476-85-1
Molecular Formula: C17H32N2O2SSn
Formula Weight: 447.22
Storage Temperature: 2°C to 8°C
Formel:
C₁₇H₃₂N₂O₂SSn Molecular Weight: 447,22 g/mol |
MDL:
MFCD07787379 CAS-Nummer: 122476-85-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...