2-Hydroxy-5-pyridinboronsäurepinakolester ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester

H51673.MD H51673.03
H51673.MDEA 24 EUR
H51673.MD H51673.03
2-Hydroxy-5-pyridinboronsäurepinakolester ≥97%
2-Hydroxy-5-pyridinboronsäurepinakolester

Purity: 97% CAS No.: 1054483-78-1 Molecular Formula: C11H16BNO3 Formula Weight: 221.06 MDL No.: MFCD06795671

Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795671
CAS-Nummer: 1054483-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR