5-Bromobenzo[c][1,2,5]oxadiazole 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2,1,3-benzoxadiazole
H51674.03
H51674.03EA
55.5
EUR
H51674.03
5-Bromobenzo[c][1,2,5]oxadiazole 97%
5-Bromobenzo[c][1,2,5]oxadiazole
Purity: 97%
CAS No.: 51376-06-8
Molecular Formula: C6H3BrN2O
Formula Weight: 199.01
MDL No.: MFCD02682026
Formel:
C₆H₃BrN₂O Molecular Weight: 199,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682026 CAS-Nummer: 51376-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...