3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific

Synonyme: tert-Butyl (2-oxo-2-((3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)amino)ethyl)carbamate, tert-Butyl (2-oxo-2-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]amino}ethyl)carbamate

H51709.06
H51709.06EA 186 EUR
H51709.06
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester 97%
3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester

Purity: 97% CAS No.: 1257651-17-4 Molecular Formula: C19H29BN2O5 Formula Weight: 376.26 MDL No.: MFCD16294500

Formel: C₁₉H₂₉BN₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD16294500
CAS-Nummer: 1257651-17-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR