Methyl-3-brom-5-hydroxybenzoat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3-bromo-5-hydroxybenzoate
H51724.03
H51724.03EA
170
EUR
H51724.03
Methyl-3-brom-5-hydroxybenzoat ≥97%
Methyl-3-brom-5-hydroxybenzoat
Purity: 97%
CAS No.: 192810-12-1
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
Melting Point: 130-135°
MDL No.: MFCD10566800
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Schmelzpunkt: 130…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10566800 CAS-Nummer: 192810-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...