4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine-1-carboxylate

H51743.03 H51743.06
H51743.03EA 72.2 EUR
H51743.03 H51743.06
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 936694-19-8 Molecular Formula: C22H35BN2O4 Formula Weight: 402.34 Beilstein Reference: 18669672 MDL No.: MFCD16294502

Formel: C₂₂H₃₅BN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16294502
CAS-Nummer: 936694-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR