4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine-1-carboxylate
H51743.03
H51743.06
H51743.03EA
72.2
EUR
H51743.03
H51743.06
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester 95%
4-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 936694-19-8
Molecular Formula: C22H35BN2O4
Formula Weight: 402.34
Beilstein Reference: 18669672
MDL No.: MFCD16294502
Formel:
C₂₂H₃₅BN₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16294502 CAS-Nummer: 936694-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...