3-Methylpyridine-2-thiocarboxamide 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H51794.03
H51794.03EA
124
EUR
H51794.03
3-Methylpyridine-2-thiocarboxamide 97%
3-Methylpyridine-2-thiocarboxamide
Purity: 97%
Molecular Formula: C7H8N2S
Formula Weight: 152.21
Melting Point: 117-122°
MDL No.: MFCD12026307
Formel:
C₇H₈N₂S Molecular Weight: 152,21 g/mol Schmelzpunkt: 117…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026307 CAS-Nummer: 334017-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...