2-Iodimidazol 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Iod-1H-imidazol

H51868.03
H51868.03EA 171 EUR
H51868.03
2-Iodimidazol 96%
2-Iodoimidazole

Purity: 96% CAS No.: 3034-62-6 Molecular Formula: C3H3IN2 Formula Weight: 193.96 Melting Point: 190-191° MDL No.: MFCD00159701

Formel: C₃H₃IN₂
Molecular Weight: 193.97 g/mol
Schmelzpunkt: 190…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159701
CAS-Nummer: 3034-62-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR