2-Chloro-1-(ethoxymethyl)imidazole 97%

Lieferant: Thermo Scientific
Achtung

H51870.03
H51870.03EA 101 EUR
H51870.03
2-Chloro-1-(ethoxymethyl)imidazole 97%
2-Chloro-1-(ethoxymethyl)imidazole

Purity: 97% CAS No.: 850429-55-9 Molecular Formula: C6H9ClN2O Formula Weight: 160.60 MDL No.: MFCD03788569

Formel: C₆H₉ClN₂O
Molecular Weight: 160,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788569
CAS-Nummer: 850429-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR