2-Chloro-4-fluoro-3-methylbenzoic acid 97%

Lieferant: Thermo Scientific
Achtung

H51879.03
H51879.03EA 203 EUR
H51879.03
2-Chloro-4-fluoro-3-methylbenzoic acid 97%
2-Chloro-4-fluoro-3-methylbenzoic acid

Purity: 97% Molecular Formula: C8H6ClFO2 Formula Weight: 188.59 Melting Point: 186-189° MDL No.: MFCD11846444

Formel: C₈H₆ClFO₂
Molecular Weight: 188,59 g/mol
Schmelzpunkt: 186…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11846444
CAS-Nummer: 173315-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR