2-(Diphenylphosphino)benzaldehyde oxime 95%
Lieferant: Thermo Scientific
|
Achtung
|
H51903.06
H51903.06EA
110
EUR
H51903.06
2-(Diphenylphosphino)benzaldehyde oxime 95%
2-(Diphenylphosphino)benzaldehyde oxime
Purity: 95%
CAS No.: 153358-05-5
Molecular Formula: C19H16NOP
Formula Weight: 305.32
MDL No.: MFCD16251585
Formel:
C₁₉H₁₆NOP Molecular Weight: 305,32 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16251585 CAS-Nummer: 153358-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...