4-(Morpholino)phenylboronsäurepinakolester 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine
H51925.03
H51925.06
H51925.03EA
65.8
EUR
H51925.03
H51925.06
4-(Morpholino)phenylboronsäurepinakolester 95%
4-(Morpholino)phenylboronsäurepinakolester
Purity: 95%
CAS No.: 568577-88-8
Molecular Formula: C16H24BNO3
Formula Weight: 289.18
MDL No.: MFCD04112544
Formel:
C₁₆H₂₄BNO₃ Molecular Weight: 289.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112544 CAS-Nummer: 568577-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...