4-(Morpholino)phenylboronsäurepinakolester 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine

H51925.03 H51925.06
H51925.03EA 65.8 EUR
H51925.03 H51925.06
4-(Morpholino)phenylboronsäurepinakolester 95%
4-(Morpholino)phenylboronsäurepinakolester

Purity: 95% CAS No.: 568577-88-8 Molecular Formula: C16H24BNO3 Formula Weight: 289.18 MDL No.: MFCD04112544

Formel: C₁₆H₂₄BNO₃
Molecular Weight: 289.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112544
CAS-Nummer: 568577-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR