9,9-Di-n-octylfluorene-2-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific

H51953.06
H51953.06EA 223 EUR
H51953.06
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester 95%
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester

Purity: 95% Molecular Formula: C35H53BO2 Formula Weight: 516.61 MDL No.: MFCD16294553

Formel: C₃₅H₅₃BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16294553
CAS-Nummer: 302554-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR