(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid
H51966.03
H51966.06
H51966.03EA
80.6
EUR
H51966.03
H51966.06
(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 98%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
Purity: 98%
CAS No.: 100564-78-1
Molecular Formula: C15H21NO4
Formula Weight: 279.34
Melting Point: 76-80°
Beilstein Reference: 3653506
Optical Rotation: +6.5° (c=2 in ethanol)
MDL No.: MFCD00076904
Formel:
C₁₅H₂₁NO₄ Molecular Weight: 279.34 g/mol Schmelzpunkt: 76…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076904 CAS-Nummer: 100564-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...