Fmoc-L-homophenylalanine 95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-HoPhe-OH, (S)-2-[[(9H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-phenylbutyric acid, Fmoc-Homophe-OH, N-Fmoc-L-homophenylalanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-homophenylalanine, (S)-2-(Fmoc-amino)-4-phenylbutyric acid

H51973.06
H51973.06EA 230 EUR
H51973.06
Fmoc-L-homophenylalanine 95%
Fmoc-L-homophenylalanine

Purity: 95% CAS No.: 132684-59-4 Molecular Formula: C25H23NO4 Formula Weight: 401.46 Beilstein Reference: 4847669 MDL No.: MFCD00077046

Formel: C₂₅H₂₃NO₄
Molecular Weight: 401,462 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077046
CAS-Nummer: 132684-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR