(R)-3-(Boc-amino)-3-phenylpropionsäure 95%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, (R)-N-BOC-3-Amino-3-phenylpropanoic acid

H51984.03
H51984.03EA 214 EUR
H51984.03
(R)-3-(Boc-amino)-3-phenylpropionsäure 95%
(R)-3-(Boc-amino)-3-phenylpropionsäure

Purity: 95% CAS No.: 161024-80-2 Molecular Formula: C14H19NO4 Formula Weight: 265.31 Beilstein Reference: 5404389 MDL No.: MFCD01320859

Formel: C₁₄H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01320859
CAS-Nummer: 161024-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR