Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-4-(Boc-aminomethyl)-L-phenylalanine

H52008.MD H52008.03
H52008.MDEA 68.8 EUR
H52008.MD H52008.03
Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%
Fmoc-4-(boc-aminomethyl)-L-phenylalanine

Purity: 95% CAS No.: 204715-91-3 Molecular Formula: C30H32N2O6 Formula Weight: 516.59 MDL No.: MFCD01317015

Formel: C₃₀H₃₂N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD01317015
CAS-Nummer: 204715-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR