Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Fmoc-4-(Boc-aminomethyl)-L-phenylalanine
H52008.MD
H52008.03
H52008.MDEA
68.8
EUR
H52008.MD
H52008.03
Fmoc-4-(boc-aminomethyl)-L-phenylalanine 95%
Fmoc-4-(boc-aminomethyl)-L-phenylalanine
Purity: 95%
CAS No.: 204715-91-3
Molecular Formula: C30H32N2O6
Formula Weight: 516.59
MDL No.: MFCD01317015
Formel:
C₃₀H₃₂N₂O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01317015 CAS-Nummer: 204715-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...