2-Benzyl-D-proline hydrochloride 95%

Lieferant: Thermo Scientific

H52019.03
H52019.03EA 417 EUR
H52019.03
2-Benzyl-D-proline hydrochloride 95%
2-Benzyl-D-proline hydrochloride

Purity: 95% CAS No.: 637020-57-6 Molecular Formula: C12H15NO2.HCl Formula Weight: 241.72 Optical Rotation: +33° (c=1 in 1N HCl) MDL No.: MFCD07367715

Formel: C₁₂H₁₅NO₂·HCl
Molecular Weight: 241,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367715
CAS-Nummer: 637020-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR