(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%

Lieferant: Thermo Scientific

Synonyme: (1R,3S)-(+)-3-Aminocyclopentane-1-carboxylic acid, N-BOC protected, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

H52022.03
H52022.03EA 315 EUR
H52022.03
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid 95%
(1S,3R)-N-Boc-1-aminocyclopentane-3-carboxylic acid

Purity: 95% CAS No.: 161660-94-2 Molecular Formula: C11H19NO4 Formula Weight: 229.28 MDL No.: MFCD01320858

Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,276 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01320858
CAS-Nummer: 161660-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR