(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

H52027.03 H52027.06
H52027.03EA 184 EUR
H52027.03 H52027.06
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 95%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid

Purity: 95% CAS No.: 734529-57-8 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 MDL No.: MFCD04113693

Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04113693
CAS-Nummer: 734529-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR