Fmoc-L-β-homophenylalanine 95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-B-HoPhe-OH, Fmoc-β-HoPhe-OH, Fmoc-β-HomoPhe-OH

H52052.03
H52052.03EA 360 EUR
H52052.03
Fmoc-L-β-homophenylalanine 95%
Fmoc-L-beta-homophenylalanine

Purity: 95% CAS No.: 193954-28-8 Molecular Formula: C25H23NO4 Formula Weight: 401.46 Beilstein Reference: 7726557 MDL No.: MFCD01863055

Formel: C₂₅H₂₃NO₄
Molecular Weight: 401,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863055
CAS-Nummer: 193954-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR