Boc-3,4-dichloro-L-phenylalanine 95%
Lieferant: Thermo Scientific
Synonyme:
Boc-Phe(3,4-DiCl)-OH
H52053.03
H52053.03EA
126
EUR
H52053.03
Boc-3,4-dichloro-L-phenylalanine 95%
Boc-3,4-dichloro-L-phenylalanine
Purity: 95%
CAS No.: 80741-39-5
Molecular Formula: C14H17Cl2NO4
Formula Weight: 334.20
Beilstein Reference: 6426101
MDL No.: MFCD00273436
Formel:
C₁₄H₁₇Cl₂NO₄ Molecular Weight: 334,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273436 CAS-Nummer: 80741-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...