Boc-D-Tyr(ME)-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

H52082.03 H52082.06
H52082.03EA 51 EUR
H52082.03 H52082.06
Boc-D-Tyr(ME)-OH 95%
Boc-D-Tyr(ME)-OH

Purity: 95% CAS No.: 68856-96-2 Molecular Formula: C15H21NO5 Formula Weight: 295.33

Formel: C₁₅H₂₁NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00065604
CAS-Nummer: 68856-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR