erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide 95%

Lieferant: Thermo Scientific

H52099.03
H52099.03EA 460 EUR
H52099.03
erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide 95%
erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide

Purity: 95% CAS No.: 388071-27-0 Molecular Formula: C15H19F2NO3 Formula Weight: 299.31 MDL No.: MFCD08061630

Formel: C₁₅H₁₉F₂NO₃
Molecular Weight: 299,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061630
CAS-Nummer: 388071-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR