Fmoc-L-Tyr(Me)-OH 95%

Lieferant: Thermo Fisher Scientific

Synonyme: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

H52117.03 H52117.06
H52117.03EA 80.2 EUR
H52117.03 H52117.06
Fmoc-L-Tyr(Me)-OH 95%
Fmoc-L-Tyr(Me)-OH

Purity: 95% CAS No.: 77128-72-4 Molecular Formula: C25H23NO5 Formula Weight: 417.45 MDL No.: MFCD00153368

Formel: C₂₅H₂₃NO₅
Molecular Weight: 417.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153368
CAS-Nummer: 77128-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR