3,3-Diphenyl-L-alanine 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: H-Ala(3,3-DiPhenyl)-OH, (S)-2-Amino-3,3-diphenylpropanoic acid

H52169.06
H52169.06EA 472 EUR
H52169.06
3,3-Diphenyl-L-alanine 95%
3,3-Diphenyl-L-alanine

Purity: 95% CAS No.: 149597-92-2 Molecular Formula: C15H15NO2 Formula Weight: 241.29 UN No.: UN2811 MDL No.: MFCD01631989

Formel: C₁₅H₁₅NO₂
Molecular Weight: 241.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631989
CAS-Nummer: 149597-92-2
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR