4-Trifluoromethyl-L-phenylalanine 95%
Lieferant: Thermo Fisher Scientific
H52170.MD
H52170.03
H52170.06
H52170.MDEA
98
EUR
H52170.MD
H52170.03
H52170.06
4-Trifluoromethyl-L-phenylalanine 95%
4-Trifluoromethyl-L-phenylalanine
Purity: 95%
CAS No.: 114926-38-4
Molecular Formula: C10H10F3NO2
Formula Weight: 233.19
Beilstein Reference: 5747857
MDL No.: MFCD00044946
Formel:
C₁₀H₁₀F₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044946 CAS-Nummer: 114926-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...