Boc-L-β-homoglutamic acid-d-benzyl ester 95%

Lieferant: Thermo Scientific

Synonyme: (S)-6-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)-6-oxohexanoic acid, Boc-β-HomoGlu(OBzl)-OH, Boc-β-HoGlu(OBzl)-OH

H52187.03
H52187.03EA 230 EUR
H52187.03
Boc-L-β-homoglutamic acid-d-benzyl ester 95%
Boc-L-beta-homoglutamic acid-d-benzyl ester

Purity: 95% CAS No.: 218943-30-7 Molecular Formula: C18H25NO6 Formula Weight: 351.40 Beilstein Reference: 8079168 MDL No.: MFCD01862936

Formel: C₁₈H₂₅NO₆
Molecular Weight: 351,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862936
CAS-Nummer: 218943-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR