(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52293.MD
H52293.03
H52293.06
H52293.MDEA
35.5
EUR
H52293.MD
H52293.03
H52293.06
(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure ≥98%
(1R,2R)-(-)-1,2-Cyclohexandicarbonsäure
Purity: 98+%
CAS No.: 46022-05-3
Molecular Formula: C8H12O4
Formula Weight: 172.18
Physical Form: Solid
Appearance: White
MDL No.: MFCD00062973
Formel:
C₈H₁₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062973 CAS-Nummer: 46022-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...