(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-Amino-3-(benzyloxy)propan-1-ol hydrochloride

H52297.06
H52297.06EA 381 EUR
H52297.06
(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride ≥98%
(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride

Purity: 98+% CAS No.: 61366-43-6 Molecular Formula: C10H15NO2.HCl Formula Weight: 217.70 MDL No.: MFCD07368914

Formel: C₁₀H₁₅NO₂·HCl
Molecular Weight: 217,70 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368914
CAS-Nummer: 61366-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR