5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%

Lieferant: Thermo Scientific

H52301.03
H52301.03EA 108 EUR
H52301.03
5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%
5'-O-(tert-Butyldimethylsilyl)thymidine

Purity: 97+% CAS No.: 40733-28-6 Molecular Formula: C16H28N2O5Si Formula Weight: 356.49 MDL No.: MFCD01631041

Formel: C₁₆H₂₈N₂O₅Si
Molecular Weight: 356,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631041
CAS-Nummer: 40733-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR