N-Benzoyl-2'-deoxyadenosine 98+%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-yl)benzamide, N6-Benzoyl-2'-deoxyadenosine, N-Benzoyl-2'-deoxy-adenosine
Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2?-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.
Purity: 98+%
CAS No.: 4546-72-9
Molecular Formula: C17H17N5O4
Formula Weight: 355.35
MDL No.: MFCD00009628
Formel:
C₁₇H₁₇N₅O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009628 CAS-Nummer: 4546-72-9 EINECS: 224-903-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...