Octahydro-2H-pyrido[1,2-a]pyrazine ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

H52305.MD H52305.03
H52305.MDEA 196 EUR
H52305.MD H52305.03
Octahydro-2H-pyrido[1,2-a]pyrazine ≥98%
Octahydro-2H-pyrido[1,2-a]pyrazine

Purity: 98+% CAS No.: 4430-75-5 Molecular Formula: C8H16N2 Formula Weight: 140.23 Boiling Point: 68-72°/3mm UN No.: UN3267 MDL No.: MFCD01098257

Formel: C₈H₁₆N₂
Siedepunkt: 68…72 °C (3 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01098257
CAS-Nummer: 4430-75-5
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR