2-Bromo-3-(chloromethyl)pyridine 96%
Lieferant: Thermo Scientific
|
Achtung
|
H52313.03
H52313.03EA
276
EUR
H52313.03
2-Bromo-3-(chloromethyl)pyridine 96%
2-Bromo-3-(chloromethyl)pyridine
Purity: 96%
CAS No.: 944906-87-0
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Storage Temperature: 2°C to 8°C
Melting Point: 34-36°
Beilstein Reference: 15651009
MDL No.: MFCD10697618
Formel:
C₆H₅BrClN Molecular Weight: 206,47 g/mol Schmelzpunkt: 34…36 °C |
MDL:
MFCD10697618 CAS-Nummer: 944906-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...