4-Bromofuran-2-carboxamide 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H52355.03
H52355.06
H52355.03EA
142
EUR
H52355.03
H52355.06
4-Bromofuran-2-carboxamide 96%
4-Bromofuran-2-carboxamide
Purity: 96%
CAS No.: 957345-95-8
Molecular Formula: C5H4BrNO
Formula Weight: 189.99
MDL No.: MFCD12755858
Formel:
C₅H₄BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12755858 CAS-Nummer: 957345-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...