2,4,5-Trimethyl-3-pyridinol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,4,5-Trimethylpyridin-3-ol

H52357.03
H52357.03EA 272 EUR
H52357.03
2,4,5-Trimethyl-3-pyridinol 97%
2,4,5-Trimethyl-3-pyridinol

Purity: 97% CAS No.: 5622-78-6 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 172-176° Beilstein Reference: 115543 MDL No.: MFCD17015035

Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17015035
CAS-Nummer: 5622-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR