6-Brom-1-tetralon 96%

Lieferant: Thermo Scientific

Synonyme: 6-Bromo-3,4-dihydro-1(2H)-naphthalenone, 6-Bromo-3,4-dihydronaphthalen-1(2H)-one

H52381.03
H52381.03EA 409 EUR
H52381.03
6-Brom-1-tetralon 96%
6-Brom-1-tetralon

Purity: 96% CAS No.: 66361-67-9 Molecular Formula: C10H9BrO Formula Weight: 225.09 Storage Temperature: 2°C to 8°C Melting Point: 40-42° MDL No.: MFCD04114378

Formel: C₁₀H₉BrO
Molecular Weight: 225,08 g/mol
Schmelzpunkt: 40…42 °C
MDL: MFCD04114378
CAS-Nummer: 66361-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR