(S)-1-Boc-2-isobutylpiperazine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-tert-Butyl 2-isobutylpiperazine-1-carboxylate, (2S)-2-Isobutylpiperazine, N1-BOC protected

H52409.MD
H52409.MDEA 139 EUR
H52409.MD
(S)-1-Boc-2-isobutylpiperazine 97%
(S)-1-Boc-2-isobutylpiperazine

Purity: 97% CAS No.: 674792-06-4 Molecular Formula: C13H26N2O2 Formula Weight: 242.36 MDL No.: MFCD03787916

Formel: C₁₃H₂₆N₂O₂
Molecular Weight: 242,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787916
CAS-Nummer: 674792-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR